Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G-Biosciences Phosphate Buffered Saline (PBS), Sterile, [10X]

Click to view available options
Quantity:
10 L
20 L
Description
- 10X PBS (Phosphate Buffered Saline) is a 10 fold concentrated, pH adjusted blend of phosphate buffer and saline solution, which when diluted 1 to 10 in ultrapure water produces a phosphate buffered saline of 8mM NaH2PO4, 150mM NaCl, 3mM KCl, and 2mM KH2PO4 at pH 7.4.
- The 10X solution is a sterile solution.
Specifications
Specifications
For Use With (Equipment) | ArcturusXT LCM Instrument |
Grade | Chemical Solution |
Product Type | Buffer Solution |
Quantity | 10 L |
Solution Type | Buffer |
Sterility | Sterile |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction