Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphoric Acid, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Catalog No. p-5391449 Shop All Avantor Products
Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
PVC Coated Glass Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.994
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
PubChem CID 1004
ChEBI CHEBI:26078
IUPAC Name phosphoric acid
SMILES OP(=O)(O)O

Specifications

Formula Weight 98
Grade BAKER ANALYZED™ ACS Reagent
Packaging PVC Coated Glass Bottle
Specific Gravity 1L = 1.69kg
Quantity 500mL
Chemical Name or Material Phosphoric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.