Learn More
VEOLIA WTS Acid Cartridge
Supplier: VEOLIA WTS APF 9031002
300-mL Acid Reagent Cartridge for the Sievers M-Series, 900 and 5310 C Series. Replacement recommended after 6 months.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.