Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphotungstic acid hydrate, 83.51%, MP Biomedicals™

Catalog No. p-4405196 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g
250 g

Chemical Identifiers

CAS 12501-23-4
Molecular Formula H5O41PW12
Molecular Weight (g/mol) 2880.20
MDL Number MFCD00011342
InChI Key AVFBYUADVDVJQL-UHFFFAOYSA-N
Synonym phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade
PubChem CID 16212977
IUPAC Name 12-Tungstophosphate hydrate
SMILES O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O

Specifications

Quantity 100 g
Physical Form Crystalline Powder
Formula Weight 2880.2
Percent Purity 83.51%
Chemical Name or Material Phosphotungstic acid hydrate
Recommended Storage Store at Room Temperature.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.