Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PHTPP, Tocris Bioscience™

Catalog No. p-4784291 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective ERβ antagonist

Selective estrogen ERβ receptor antagonist that displays 36-fold selectivity over ERα. Exhibits full antagonism at ERβ in a cotransfection assay in human endometrial cancer cells (HEC-1), with minimal effects on ERα.

Chemical Identifiers

CAS 805239-56-9
Molecular Formula C20H11F6N3O
Molecular Weight (g/mol) 423.318
InChI Key AEZPAUSGTAHLOQ-UHFFFAOYSA-N
Synonym phtpp, 4-2-phenyl-5,7-bis trifluoromethyl pyrazolo 1,5-a pyrimidin-3-yl phenol, d02wru, pyrazolo 1,5-a pyrimidine, 2a, phtpp hplc
PubChem CID 11201035
IUPAC Name 4-[2-phenyl-5,7-bis(trifluoromethyl)pyrazolo[1,5-a]pyrimidin-3-yl]phenol
SMILES C1=CC=C(C=C1)C2=NN3C(=CC(=NC3=C2C4=CC=C(C=C4)O)C(F)(F)F)C(F)(F)F

Specifications

Quantity 10 mg
Formula Weight 423.31
Percent Purity >99%
Chemical Name or Material PHTPP
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.