Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
phyproof™ Salvianolic acid B, ≥90% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
:
10 mg
Specifications
Specifications
CAS | 121521-90-2 |
Quantity | 10 mg |
Type | Reference Substance |
Product Line | phyproof |
Linear Formula | C36H30O16 |
MDL Number | MFCD23115783 |
Synonym | (2 S,3 S)-4-[(1 E)-3-[(1 R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy]-3-oxo-1-propen-1-yl]-2-(3,4-dihydroxyphenyl)-2,3-dihydro-7-hydroxy-3-benzofurancarboxylic acid 3-[(1 R)-1-carboxy-2-(3,4-dihydroxyphenyl)ethyl] ester; Dan Shen Suan B; Danfensuan B; Lithospermic acid B |
Formula Weight | 718.61 |
Percent Purity | ≥90% (HPLC) |
Grade | Primary Reference Standard |
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction