Learn More
Invitrogen™ PichiaPink™ Secreted Vector Kit

The PichiaPink™ Secreted Vector Kit comes with:
Why Choose the PichiaPink™ Yeast Expression System?
The PichiaPink™ Yeast Expression System is based on the yeast Pichia pastoris. Advantages of Pichia pastoris include rapid growth, well-defined genetic background, simple media formulation, and easy handling. For over 30 years, Pichia pastoris has been used by labs around the world for producing hundreds of different proteins from many species including human (Ref 1, 2). The PichiaPink™ Yeast Expression System allows convenient and cost-effective protein production from small to large scales.
For information on obtaining a commercial-use license for the PichiaPink™Yeast Expression System, please inquire at outlicensing@lifetech.com.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Related Links
References
1. Cereghino JL, Cregg JM. Heterologous protein expression in the methylotrophic yeast Pichia pastoris. FEMS Microbiol Rev. 2000 Jan;24(1):45-66. [PubMed]
2. Cereghino GP, Cereghino JL, Ilgen C, Cregg JM. Production of recombinant proteins in fermenter cultures of the yeast Pichia pastoris. Curr Opin Biotechnol. 2002 Aug;13(4):329-32. [PubMed]
Specifications
Format
Liquid
Inducing Agent
Methanol
Promoter
AOX1
Product Type
Vector Kit
Content And Storage
PichiaPink™ Secreted Protein Vector kit contains 20 μg pPinkα-HC (the high-copy number plasmid with the S. cerevisiae Alpha mating factor presequence) with sequencing primers 5' α-factor and 3' CYC1. Store at -20°C.
Antibiotic Resistance Bacterial
Ampicillin (AmpR)
Cleavage
No Cleavage Site
Replication Origin
pUC Origin
Shipping Condition
Room Temperature
Protein Tag
Untagged
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.