Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ MS(PEG)8 Methyl-PEG-NHS-Ester Reagent
Shop All Thermo Scientific Products
Click to view available options
:
8 PEG groups
Description
Thermo Scientific™ Pierce MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) activated as NHS esters for covalent pegylation of primary amines on proteins (e.g., lysines) or assay surfaces.
MS(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with methyl (—CH3) and amine-reactive NHS-ester groups at opposite ends. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-NHS Ester, where the subscript “n” denotes 4, 8, 12, or 24 ethylene glycol units. The N-hydroxysuccinimide (NHS) ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.
Highlights:
- NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
- Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
- Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Easy-to-follow instructions increase the likelihood of a successful outcome
Specifications
Specifications
Chemical Reactivity | Amine |
Description | Pierce MS(PEG)8 |
Content And Storage | Store desiccated at -20°C. |
Label Type | Polyethylene Glycol (PEG), Blocking Groups |
Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
Spacer | 8 ethylene glycol units |
Form | Liquid |
Quantity | 100 mg |
Solubility | Water |
Product Type | NHS-Ester Reagent |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction