Learn More
Medchemexpress LLC HY-B1101A 10mg Medchemexpress, Pimethixene maleate CAS:13187-06-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1101A10MG
Encompass_Preferred
Medchemexpress, HY-B1101A 10mg Pimethixene maleate CAS:13187-06-9 Pimethixene maleate is antihistamine and antiserotonergic compound, acts as an antimigraine agent.Pimethixene maleate is a highly potent antagonist of 5-HT1A, 5-HT2A, 5-HT2B, 5-HT2C, histamine H1, dopamine D2 and D4.4 as well as muscarinic M1 and M2 receptors, with pKis of 7.63, 10.22, 10.44, 8.42, 10.14, 8.19, 7.54, 8.61 and 9.38, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0139
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.