Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pimozide, Tocris Bioscience™

Catalog No. 0937100 Shop All R&D Systems Products
Click to view available options
Quantity:
This item is not returnable. View return policy
This item is not returnable. View return policy

D2-like antagonist

Dopamine D2-receptor antagonist and antipsychotic which binds with very high affinity to the cloned rat 5-HT7 receptor (Ki = 0.5 nM).

Chemical Identifiers

CAS

2062-78-4

Molecular Formula

C28H29F2N3O

Molecular Weight (g/mol)

461.56

MDL Number

MFCD00055081

InChI Key

YVUQSNJEYSNKRX-UHFFFAOYSA-N

Synonym

pimozide, orap, opiran, pimozidum, pimozidum inn-latin, mcn-jr-6238, pimozida, primozida, primozida inn-spanish, primozide

PubChem CID

16362

ChEBI

CHEBI:8212

IUPAC Name

1-{1-[4,4-bis(4-fluorophenyl)butyl]piperidin-4-yl}-2,3-dihydro-1H-1,3-benzodiazol-2-one

SMILES

FC1=CC=C(C=C1)C(CCCN1CCC(CC1)N1C(=O)NC2=CC=CC=C12)C1=CC=C(F)C=C1

Specifications

Quantity

100 mg

Formula Weight

461.55

Percent Purity

>99%

Chemical Name or Material

Pimozide

Recommended Storage

Store at Room Temperature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.