Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ CleanTeam™ Seamless Knit Nylon/Copper Fiber ESD Gloves with Polyurethane Finger Grip
GSA_VA

Catalog No. p-3348671 Shop All Protective Industrial Products Inc. Products
Click to view available options
Size:
Small
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Electrostatic Dissipative (ESD) fiber yarns blended with low lint nylon reduce static build-up on the glove's surface for improved performance in electronics assembly. Polyurethane coating on the fingertips provides a secure grip in dry or slightly oily conditions.

  • Electrostatic Dissipative (ESD) fiber yarns blended with low lint nylon
  • Reduced static build-up on the glove's surface for improved performance in electronics assembly
  • Polyurethane coating on the fingertips enhances grip in dry or slightly oily conditions
  • Electrical Surface Resistivity (ASTM D 257:2007): Below 7.5 x 107
  • Seamless knit construction enhances dexterity and tactility

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Coated Area Fingertips
Construction Seamless Knit
Coating Material Polyurethane
Color Natural
Disposable Reusable
Exterior Finish No Texture
Material Nylon, Copper Fiber
Product Type ESD Gloves
Size Small
For Use With (Application) Aerospace, Circuit Board Manufacturing, Electronics Assembly, and Munitions
Glove Cuff Style Knit
Hem Color Red
Series 40-6416
Yarn Gauge 15
Length (English) 8.3 in.
Length (Metric) 21 cm
Show More Show Less

WARNING: Cancer - www.P65Warnings.ca.gov

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.