Learn More
TARGETMOL CHEMICALS INC PIPERLONGUMINE 100MG
Supplier: TARGETMOL CHEMICALS INC T6947
Also available in 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Piperlongumine (Piplartine) is a natural alkaloid from Piper longum L. It selectively kills cancer cells and strengthens the level of reactive oxygen species (ROS). purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.