Learn More
Research Products International Corp Pipet Box Organizer, Horizontal, 4 Box Capacity of 16 1/2 Inch Long Pipet Boxes
SDP
Supplier:  Research Products International Corp 145621
Encompass
Clear acrylic organizers accommodate square and round stainless steel pipet boxes. Alllows box cover to be removed with one hand operation. To prevent possible warping of organizers, allow pipet cans to cool to room temperature before placing in racks. Capacity four boxes.
Catalog No. 50-212-322
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.