Learn More
Research Products International Corp Pipet Sterilizing Can, Stainless Steel, Square, 2 x 2 x 9 1/4 Inches
SDP
Supplier:  Research Products International Corp 1369
Encompass
Pipet sterilizing cans made from type 304 stainless steel offer high resistance to corrosion.
Catalog No. 50-212-141
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.