Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Roylco™ Planet Earth Craft Paper
SCI_ED

Catalog No. S13224
Click to view available options
:

Celebrate the only planet that we can call our home with this huge collection of illustrated craft papers inspired by the environments of our beautiful Earth.

  • Each sheet is full of colorful, detailed subjects ranging from grass and stones to coniferous forests and seascapes
  • Use them for building your own mixed media collage art, covering your decorative nature journal, accents, scrapbooks, paper sculptures and more
  • Ideal for Earth Day craft activities
  • 24 different designs
  • Dimensions: 8.5 x 5.5 in. (21.6 x 14 cm)

Specifications

Grade

1+

Format

Kit

No. of Pages

144 Pages

Languages

English, Spanish, French, German

Subject

Earth Science

Length (English)

8.5 in.

Length (Metric)

21.6 cm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.