Learn More
Electron Microscopy Sciences Evactron Zephyr Plasma Decontaminator
Supplier:  Electron Microscopy Sciences 9100010
Encompass_Preferred
Evactron Model 25 Zephyr Plasma Decontaminator is for SEM/FIB systems that use turbo molecular pumps. IT provides fast and efficient hydrocarbon removal using a remote RF plasma to produce gas-phase radicals that flow downstream through the chamber. There is no damage to samples or sensitive components. It cleans/de-contaminates in the turbo pressure (TMP) regime at 1-50 mTorr and has no adverse effects on the TMP temperatures. It features a desktop controller, SEM/FIB chambers or load locks, and 2 operating regimes-classic mode (roughing pressures) and T-pump mode (turbomolecular pressures).
Catalog No. 50-192-8218
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.