Learn More
Epoch Life Science Inc GenCatch™ Plasmid DNA Midiprep Kit (50 preps) Uniquely formulated buffers, reinforced anion exchange columns with collection tubes, along with protocols.
SDP
Supplier:  Epoch Life Science Inc 2170050
Encompass_Preferred
Provides a simple, fast and cost-effective method to purify transfection grade plasmid DNA using ion (anion) exchange technology without phenol extraction or ethanol precipitation. 100 ug plasmid DNA binding capacity. Gravity flow. DNA prepared by this kit is suitable for many downstream molecular manipulations including PCR, ligation, restriction enzyme digestion, transformation and transfection, radioactive and fluorescent sequencingg, etc.
Catalog No. 50-153-8114
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.