Learn More
Innovative Med Tech Plastic Blood Gas Capillary Tubes: Additives
Additives for Innovative Med Tech Plastic Blood Gas Capillary Tubes
Supplier:  Innovative Med Tech 102065
 View more versions of this product
Catalog No. 22-757-140
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Description
Description
  • Made from 100% plastic
  • Use for blood gas testing, analyte testing, fetal scalp pH tests, and newborn screening
Compliance
Meets FDA, OSHA, and CDC mandates
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.