Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Plastic Slide Grip, Plastic Slide Grip

Catalog No. 1975 Shop All Epredia Products
Click to view available options
Product Type:

Ensure safety in slide handling during staining with the Epredia™ Plastic Slide Grip.

  • Holds five (76 x 26mm) slides
  • Can be used in temperatures to 80°C

Specifications

Product Type

Plastic Slide Grip

Holds

Slides

Material

Plastic

Positions

5

Length (Metric) Slide

76 mm

Width (Metric) Slide

26 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.