Learn More
Cole-Parmer Argos Technologies Sterile Tissue Culture 6-Well Plate, 100/cs
Supplier:  Cole-Parmer 0192770
Encompass_Preferred
Use these culture well plates to serve your various cell and tissue culture needs. The plates are manufactured in a cleanroom environment for optimal quality. They have treated surfaces of an exclusive coating, which causes the surface to become negatively charged and hydrophilic, allowing for consistent and superior cell attachment and growth. They also have individual condensation rings, and a one-way fitting lid to reduce cross-contamination. The alphanumeric notation on each well allows for simple identification.
Catalog No. 50-196-0585
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.