Learn More
Strem, An Ascensus Company CAS# 7440-06-4. 5g. Platinum, 5% on calcium carbonate, unreduced, dry (Escat™ 2371). MFCD00011179
SDP
Supplier:  Strem, An Ascensus Company 7816655G
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CAS# 7440-06-4. 5g. Platinum, 5% on calcium carbonate, unreduced, dry (Escat™ 2371). MFCD00011179. Molecular Weight: . Molecular Formula: . Color/form: black pwdr. (d50=3 mum) Strem# 78-1665. http://www.strem.com/catalog/v/78-1665/
Catalog No. 50-901-13282
missing translation for 'QuantityLeft'