Learn More
Glass Expansion Platinum Skimmer Cone Ni
Supplier: Glass Expansion AT8908SPT/NI
Platinum Skimmer Cone with Nickel Base for s-lens (G3666-67411). For Agilent 8900 ICP-MS. Recommended for organic solvents and added oxygen (e.g., volatile and semi-volatile solvents). More durable, longer-lasting, & because it runs hotter it requires less frequent cleaning. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. Use Skimmer ConeGuard (70-803-1004) to protect cone threads during cleaning. When not in use, keep cones stored in original packaging for protection.For routine applications, use standard Nickel Skimmer for x-lens, see P/N 501907593.For organic analysis, use Platinum Skimmer with Ni Base for x-lens, P/N 501907595, which must be mounted on brass adaptor skimmer base, see P/N AT7905X-BR. For lowest nickel background and laser ablation applications, use Aluminum Skimmer for x-lens, see P/N AT7902X-Al.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.