Learn More
Mirus Bio pLIVE™ Vector/lacZ Control Vector Kit
pLIVE In Vivo Expression and Reporter Vectors is a vector system designed to achieve long term expression in the livers of laboratory mice.
Supplier: Mirus Bio MIR 5520
This item is not returnable.
View return policy
Description
- Sustained Gene Expression: Achieve long term liver gene expression in the mouse liver through 8 months post-injection
- Versatile Platform: Available with positive control vectors expressing SEAP