Learn More
Chemglass Life Sciences Plug, Drosophila, High Density Foam, for 6oz Bottles
Supplier: Chemglass Life Sciences CGE4790005
• Fly plugs are made of high-density foam• Causes minimal shedding and reduce evaporation• Maintains tight fit after repeated use and removal• Plugs are compatible with CGE-4789 Drosophila vials and bottles (Sold Separately)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.