Learn More
Pluriselect PLURISPIN HUMAN PLT DEPLETION
SDP
Supplier:  Pluriselect 190000231
Encompass_Preferred
pluriSpin Human PLT Depletion is designed to isolate pure, viable and untouched platelets-low population from whole blood, buffy coat, cord blood or similar sample material by negative isolation. The product labels Platelets, while the isolated human platelets-low population are left in the sample. When centrifuged over a density medium. The unwanted cells pellet along with the RBCs. The purified platelets-free population are present as a highly enriched population at the interface between the plasma and the density medium.
Catalog No. NC2171785
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.