Learn More
ILEX LIFE SCIENCES LLC PLURONIC F-68 10 SOL 6X100ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0802100X6
Encompass_Preferred
PAN-Biotech Pluronic F-68 10% Solution (6 x 100 ml). DESCRIPTION: Pluronic F-68 is a difunctional block copolymer surfactant terminating in primary hydroxyl groups. It is a non-ionic detergent that protects cells from hydrodynamic damage. It can be also used for in vitro isolation of lymphocytes from diluted whole blood. APPLICATION: Pluronic F-68 is a non-ionic surfactant in cell culture used to control shear forces in suspension cultures against possible damage during freezing and thawing, transfer and stirring. It can also be used to reduce foaming in stirred cultures and reduce cell attachment to glass. Pluronic F-68 prevents agglomeration of cells. It can be also used for in vitro isolation of lymphocytes from diluted whole blood. CAS REGISTRY NO.: 9003-11-6. LIQUID/POWDER: Liquid. STERILE: Yes. STORAGE TEMP: Room Temperate (RT). MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3702
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.