Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMPA (NAALADase inhibitor), Tocris Bioscience™

Catalog No. p-4785362 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GCP II inhibitor

Highly potent and selective inhibitor of glutamate carboxypeptidase 2 (GCP II/N-acetylated α-linked dipeptidase/NAALADase) with a Ki value of 275 pM. Neuroprotective; protects against glutamate-mediated motor neuron degeneration and reduces volume of injury following middle cerebral artery occlusion (MCAO).

Chemical Identifiers

CAS 173039-10-6
Molecular Formula C6H11O7P
Molecular Weight (g/mol) 226.121
InChI Key ISEYJGQFXSTPMQ-UHFFFAOYSA-N
Synonym 2-phosphonomethyl pentanedioic acid, 2-pmpa, pmpa naaladase inhibitor, 2-phosphonomethyl-pentanedioic acid, chembl47009, pmpa naaladaseinhibitor, d0f3zo, d0y6ub, 2-pmpa naaladaseinhibitor
PubChem CID 10130754
IUPAC Name 2-(phosphonomethyl)pentanedioic acid
SMILES C(CC(=O)O)C(CP(=O)(O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 226.12
Percent Purity >98%
Chemical Name or Material PMPA (NAALADase inhibitor)
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.