Learn More
Gold Biotechnology Inc Phenylmethylsulfonyl fluoride (PMSF), 329-98-6, MFCD00007424, 10 g
SDP
Supplier:  Gold Biotechnology Inc P47010
Encompass_Preferred
Molecular Formula: C7 H7 F O2 S, Molecular Weight: 174.194, Store desiccated at 4°C, White to slightly off white cyrstalline powder.
Catalog No. 50-153-2823
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.