Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PNU 37883 hydrochloride, Tocris Bioscience™

Catalog No. p-4784323 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Vascular Kir6 (KATP) channel blocker

Novel antagonist selective for the vascular form of Kir6 (KATP) channel; inhibits Kir6 currents in isolated mesenteric artery smooth muscle cells (Kd = 65 nM) but not in cardiac or skeletal myocytes. Inhibits blood vessel relaxation and hypotension induced by pinacidil in vivo. Displays weak diuretic effects.

Chemical Identifiers

CAS 57568-80-6
Molecular Formula C21H36ClN3O
Molecular Weight (g/mol) 381.989
InChI Key FZALCKUJYZCDOX-UHFFFAOYSA-N
Synonym pnu 37883 hydrochloride, 4-morpholinecarboximidine-n-1-adamantyl-n'-cyclohexane, n-cyclohexyl-n'-tricyclo 3.3.1.1 3,7 dec-1-yl-4-morpholinecarboximidamide monohydrochloride, 4-morpholinecarboximidamide, n-cyclohexyl-n'-tricyclo 3.3.1.1 3,7 dec-1-yl-, monohydrochloride, c21h35n3o.hcl, n-1-adamantyl-n'-cyclohexylmorpholine-4-carboximidamide hydrochloride, n-cyclohexyl-n'-tricyclo 3.3.1.13,7 dec-1-yl-4-morpholinecarboximidamide hydrochloride, n-1-adamantyl-n inverted exclamation marka-cyclohexyl-4-morpholinecarboxamidine hydrochloride
PubChem CID 64392
IUPAC Name N-(1-adamantyl)-N'-cyclohexylmorpholine-4-carboximidamide;hydrochloride
SMILES C1CCC(CC1)N=C(NC23CC4CC(C2)CC(C4)C3)N5CCOCC5.Cl

Specifications

Quantity 10 mg
Formula Weight 381.98
Percent Purity >99%
Chemical Name or Material PNU 37883 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.