Learn More
TARGETMOL CHEMICALS INC PNU-96391 5MG
Supplier:  TARGETMOL CHEMICALS INC T28435
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PNU-96391 (OSU-6162) is an antagonist of dopamine D2 and a partial agonist of 5-HT2A. PNU-96391 can be used in research on the treatment of chronic diseases. purity: 99%
Catalog No. 50-258-0517
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.