Learn More
Electron Microscopy Sciences Pikal Polishing Paste
Supplier:  Electron Microscopy Sciences 7183102
Encompass_Preferred
A thick abrasive paste for cleaning and suface finishing. Good for removing stains on enamel, glass and metal. Can be applied to cotton swabs or wipes for polishing. Test first before using on soft metals.
Catalog No. 50-192-8695
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.