Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poly(Bisphenol A carbonate), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
:
25 g
Description
Poly(Bisphenol A carbonate) (PC) is a thermoplastic material. It is injected in molding operations and is majorly studied to understand its thermal behaviour, especially thermal degradation property. It can be synthesized by inducing supercritical CO2 to crystallinity in low molecular weight polycarbonate beads using solid-state polymerization method.
Specifications
Specifications
CAS | 25037-45-0 |
Quantity | 25 g |
Type | Analytical Standard |
Purity Grade Notes | A lot specific analysis is included with each unit |
MDL Number | MFCD00084476 |
Grade | Analytical Standard |
Density | 1.2 g/mL (at 25°C) |
Physical Form | Neat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction