Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PolyCo™ VR™ Rainwear Overalls
Click to view available options
Size:
Medium
Large
X-Large
Specifications
Specifications
Closure Type | Storm Flap with Heavy-duty Buckles |
Color | Yellow |
Material | PVC |
Size | Large |
Construction | 3-piece |
For Use With (Application) | Impervious to fats and Oils |
Thickness | 8 mil |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction