Learn More
Polyethylene Glycol 40 Castor Oil, Hydrogenated, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Specifications
Color
Yellow liquid
Acid Value
3
CAS Max %
1
Quantity
470 lb.
Grade
Reagent
Hydroxyl Value
59-68
Packaging
Poly Drum
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.