Learn More
Microsolv Technology Corporation Polymer Tubing Drill. 0.8mm Polymeric Pin, Replacement for Flanging Tool. 1 EA.
Supplier:  Microsolv Technology Corporation 45099108
Encompass_Preferred
This Drill is designed to flange the tips of Polymer (plastic) tubing when used with the MicroSolv Flanging Tool.
Catalog No. 50-227-1419
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.