Learn More
Microsolv Technology Corporation Polymer Tubing Drill. 1.5mm Polymeric Pin, Accessory for Flanging Tool. 1 EA.
Supplier: Microsolv Technology Corporation 45099115
This Drill is designed to flange the tips of Polymer (plastic) tubing when used with the MicroSolv Flanging Tool and is not included with the tool or with the kit.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.