Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 20, NF, Spectrum™ Chemical
SDP

Catalog No. p-7158028 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum

PO132, 9005-64-5

Spectrum™ Chemical Polysorbate 20, NF is a hydrophilic, nonionic, polysorbate surfactant and a polyoxyethylene derivative of sorbitan monolaurate with relative non-toxicity. It is used in cosmetic and personal care product formulations as a fragrance ingredient, surfactant, emulsifier, and wetting agent and is used in a variety of products including skin fresheners, skin care products, skin cleansing products, makeup bases and foundations, shampoos, permanent waves and fragrance powders. It appears as a yellow liquid and is soluble in cold water. Spectrum Chemical NF products, including all NF polysorbate products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Acid Value 2
CAS Max % 1
Quantity 6 x 500 mL
Hydroxyl Value 96-108
Ignition Residue 0.0025
Packaging Amber Glass Bottle
Saponification Value 40-50
Water 0.03
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.