Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 60, NF, Spectrum™ Chemical
SDP

Catalog No. p-7158031 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 mL
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum

PO136, 9005-67-8

Spectrum™ Chemical Polysorbate 60, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9005-67-8
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC24H46O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD01780110
InChI Key DEKFZWMENCCOFU-UHFFFAOYNA-N
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Acid Value 2
CAS Max % 1
Quantity 500 mL
Hydroxyl Value 81-96
Ignition Residue 0.0025
Packaging Amber Glass Bottle
Saponification Value 45-55
Water 0.03
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.