Learn More
United Scientific Supplies Porcelain Mortar and Pestle Set



Porcelain Mortar and Pestle Set is neatly glazed for a consistent finish.
Supplier: United Scientific Supplies JMD400
Description
- Deep form mortars with lips are glazed on the outside (except for the bottom) and unglazed inside.
- Oversized pestles are glazed to grinding surface.
- Includes both mortars and pestles
- Available in 7 sizes ranging from 50 mL to 1900 mL.
Specifications
Glazed Outer, Unglazed Inner | |
400 mL | |
Mortar and Pestle | |
Porcelain |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.