Learn More
Brady™ Portable Metal Lock Box, Spanish
Shop All Brady Corporation Products- For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
- Reduces the number of locks being used
- Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
- Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Color
Red
Dimensions (L x W x H)
3.5 x 9 x 6 in.
Material
Powder-coated Steel
Type
Portable Lock Box
Displayed Text
CAJA DE TRABAS TRABADO DE SEGURIDAD
Height (English)
6 in.
Length (English)
3.5 in.
Quantity
1 Ea.
Width (English)
9 in.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.