Learn More
Allows safe and easy transport of equipment from countertop to cart.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type
Equipment Cart
Dimensions (L x W x H)
34.88 x 19.0 x 36.38 in. (88.6 x 48.3 x 92.4 cm)
Includes
Two open shelves; 4 in. diameter casters, two with toe locks; Extruded rubber handle grips; Glacier white, epoxy-coated 18 gauge welded tubular steel frame; ID tag
Load Bearing Capacity (English)
400 lb.
Load Bearing Capacity (Metric)
181 kg
Material
Steel
No. of Casters
4
No. of Shelves
2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.