Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Benzoate, NF, 99-101%, Spectrum™ Chemical
SDP

Catalog No. p-7158045 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

P1181, 582-25-2, C7H5KO2

Spectrum™ Chemical Potassium Benzoate, NF is used as a preservative that limits the growth of yeast, mold and some bacteria. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 582-25-2
Molecular Formula C7H5KO2
Molecular Weight (g/mol) 160.21
InChI Key XAEFZNCEHLXOMS-UHFFFAOYSA-M
IUPAC Name potassium benzoate
SMILES [K+].[O-]C(=O)C1=CC=CC=C1

Specifications

Alkalinity To pass test
CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99 to 101%
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.