Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Carbonate, Sesquihydrate
SCI_ED

Catalog No. S25479 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
500 g
Packaging:
Solid

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 6381-79-9
Molecular Formula C2H6K4O9
Molecular Weight (g/mol) 330.45
MDL Number MFCD00149925
InChI Key WWVGGBSGYNQKCY-UHFFFAOYSA-J
Synonym Pearl Ash, Pearl Ash Sesquihydrate
IUPAC Name tetrapotassium trihydrate dicarbonate
SMILES O.O.O.[K+].[K+].[K+].[K+].[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Quantity 500 g
Assay Percent Range 0.985
Linear Formula K2CO3 ·1.5H2O
Formula Weight 165.24
Packaging Solid
Density 2.155g/mL
Chemical Name or Material Potassium Carbonate, Sesquihydrate
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.