Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Potassium Channel Kv2.2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB300261
Description
Potassium Channel Kv2.2 Polyclonal antibody specifically detects Potassium Channel Kv2.2 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Potassium Channel Kv2.2 | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:1000, Immunohistochemistry-Frozen 1:10-1:500 | |
Q63099 | |
KCNB2 | |
Synthetic peptide corresponding to amino acid residues specific to the Kv2.2 subunit conjugated to KLH. Accession # Q63099 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼125k voltage gated potassium channel, Kv 2.2 subunit. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
delayed rectifier potassium channel protein, KV2.2, potassium channel Kv2.2, potassium voltage-gated channel subfamily B member 2, potassium voltage-gated channel, Shab-related subfamily, member 2, voltage-gated potassium channel subunit Kv2.2 | |
Rabbit | |
100 kDa | |
0.1 mL | |
Neuronal Cell Markers, Neuroscience, Neurotransmission, Potassium Channels | |
9312 | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction