Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Chloride Conductivity Standard, 5 μS/cm at 25°C, Ricca Chemical
SDP

Catalog No. p-4770373 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 7447-40-7, 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Packaging Natural Poly Bottle
Quantity 500 mL
Color Colorless
Assay Percent Range 5.00 ± 0.03 μS/cm at 25°C
Odor Odorless
Solvent or Matrix Aqueous Solution
Solubility Infinitely soluble in water
Physical Form Liquid
Chemical Name or Material Potassium Chloride Conductivity Standard, 5μS/cm (5μmho/cm) at 25°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.