Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Potassium Ferricyanide
SCI_ED

Catalog No. p-4416428 Shop All AquaPhoenix Scientific Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Solid
This item is not returnable. View return policy
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Orange to Red
Quantity 500 g
Assay Percent Range >95%
Linear Formula K3Fe(CN)6
Formula Weight 329.26
Packaging Solid
Chemical Name or Material Potassium Ferricyanide
ShelfLife 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.