Learn More
Strem, An Ascensus Company CAS# 18583-60-3. 1g. Potassium hydrotris(pyrazol-1-yl)borate hydrate, 98%. MFCD03414149
SDP
Supplier:  Strem, An Ascensus Company 1936001G
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
CAS# 18583-60-3. 1g. Potassium hydrotris(pyrazol-1-yl)borate hydrate, 98%. MFCD03414149. Molecular Weight: 252.13. Molecular Formula: [C9H10BN6]K∙XH2O. Color/form: white pwdr. Strem# 19-3600. http://www.strem.com/catalog/v/19-3600/
Catalog No. 50-901-10643