Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Hydroxide, 50% (w/v), Suitable for Orsat Gas Analysis, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
55 gal.
Packaging:
Natural Poly Bottle
Poly Drum
Description
Suitable for Orsat Gas Analysis
Specifications
Specifications
Quantity | 500 mL |
Formula Weight | 56.11 |
Concentration or Composition (by Analyte or Components) | 50% (w/v) Aqueous Solution |
Packaging | Natural Poly Bottle |
Chemical Name or Material | Potassium Hydroxide Solution, 50% (w/v) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction