Learn More
Hach Company Potassium Hydroxide Standard Solution, 8.00 N, 500 mL
Supplier:  Hach Company 28249
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
For pH adjustment in calcium determination by EDTA titration. Hach methods 8222 (buret) and 8204 (Digital Titrator). 500 mL bottle.
Catalog No. 50-199-4233