Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium ICP Standard, 10,000 ppm K in 5% HNO3, Ricca Chemical
SDP

Catalog No. p-3252171 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle

Chemical Identifiers

CAS 7757-79-1, 7697-37-2, 7732-18-5
Molecular Formula KNO3
Molecular Weight (g/mol) 101.10
MDL Number MFCD00011409
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
PubChem CID 24434
ChEBI CHEBI:63043
IUPAC Name potassium nitrate
SMILES [K+].[O-][N+]([O-])=O

Specifications

Packaging Natural Poly Bottle
Quantity 50 mL
CAS Min % 89.0, 6.69, 2.3
CAS Max % 93.0, 6.96, 2.4
Color Colorless
Concentration or Composition Notes 10,000ppm K
Equivalence 1 mL = 10 mg K
Physical Form Liquid
Chemical Name or Material Potassium ICP Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.